AMBH324A4DA8 Ambeed, Inc.

(2S,3R,4S,5R)-2,3,4,5-Tetrahydroxy-6-oxohexanoic acid hydrate(mixture of isomers)

  • Cas Number : 91510-62-2
  • Molecular Weight : 212.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/91510-62-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/91510-62-2.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart