AMBH324A4DA9 Ambeed, Inc.

3,4,5-Trifluoro-4'-[(trans,trans)-4'-pentyl[1,1'-bicyclohexyl]-4-yl]-1,1'-biphenyl

  • MDL number MFCD11053475
  • Cas Number : 137529-43-2
  • Molecular Weight : 442.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/137529-43-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/137529-43-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart