AMBH324A4DAA Ambeed, Inc.

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)non-8-enoic acid

  • MDL number MFCD19381837
  • Cas Number : 1262886-63-4
  • Molecular Weight : 393.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1262886-63-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1262886-63-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart