AMBH324A4DBE Ambeed, Inc.

(R)-2-(Piperidin-3-yl)isoindoline-1,3-dione hydrochloride

  • MDL number MFCD09752972
  • Cas Number : 1381795-31-8
  • Molecular Weight : 266.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1381795-31-8.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1381795-31-8.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart