AMBH324A4DC5 Ambeed, Inc.

(2S,4R)-1-tert-Butyl 2-ethyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride

  • MDL number MFCD28411204
  • Cas Number : 2187426-87-3
  • Molecular Weight : 294.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2187426-87-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2187426-87-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart