AMBH324A4DCE Ambeed, Inc.

(E)-4-Phenylbut-3-en-2-one

  • MDL number MFCD00008779
  • Cas Number : 1896-62-4
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1896-62-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1896-62-4.html
pictograms GHS07
hstatements H315-H317-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart