AMBH324A4DE6 Ambeed, Inc.

(S)-3-Amino-4-(3-chlorophenyl)butanoic acid

  • MDL number MFCD08461687
  • Cas Number : 270596-38-8
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/270596-38-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/270596-38-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart