AMBH324A4E04 Ambeed, Inc.

4-(trans-4-n-Propylcyclohexyl)benzoic acid

  • MDL number MFCD06658177
  • Cas Number : 65355-29-5
  • Molecular Weight : 246.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/65355-29-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/65355-29-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart