AMBH324A4E05 Ambeed, Inc.

(R)-Methyl 2-((tert-butyldimethylsilyl)oxy)propanoate

  • MDL number MFCD28402584
  • Cas Number : 171230-81-2
  • Molecular Weight : 218.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/171230-81-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/171230-81-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart