AMBH324A4E15 Ambeed, Inc.

Fmoc-D-Pra-OH

  • MDL number MFCD01311781
  • Cas Number : 220497-98-3
  • Molecular Weight : 335.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/220497-98-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/220497-98-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart