AMBH324A4E1A Ambeed, Inc.

Fmoc-Phe(3-F)-OH

  • MDL number MFCD00672328
  • Cas Number : 198560-68-8
  • Molecular Weight : 405.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/198560-68-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/198560-68-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart