AMBH324A4E28 Ambeed, Inc.

Cryptotanshinone

  • MDL number MFCD07636810
  • Cas Number : 35825-57-1
  • Molecular Weight : 296.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/35825-57-1.html

Safety Information

signalword Danger
pcodes P273-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/35825-57-1.html
pictograms GHS06,GHS09
hstatements H301-H410
Availability details for
Quantity
  • Availability
  • Add to Cart