AMBH324A4E2D Ambeed, Inc.

(Z/E-)-Polydatin

  • MDL number N/A
  • Cas Number : 65914-17-2
  • Molecular Weight : 390.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/65914-17-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/65914-17-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart