AMBH324A4E2E Ambeed, Inc.

(trans,trans)-4-(2,3-Difluoro-4-methylphenyl)-4'-ethyl-1,1'-bi(cyclohexane)

  • MDL number N/A
  • Cas Number : 174350-08-4
  • Molecular Weight : 320.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/174350-08-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/174350-08-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart