AMBH324A4E30 Ambeed, Inc.

(2E,4E)-Octa-2,4-dienal

  • MDL number MFCD00014676
  • Cas Number : 30361-28-5
  • Molecular Weight : 124.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/30361-28-5.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/30361-28-5.html
pictograms GHS07
hstatements H227-H315
Availability details for
Quantity
  • Availability
  • Add to Cart