AMBH324A4E34 Ambeed, Inc.

(2S,5R,6R)-6-(3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid,sodium salt

  • MDL number MFCD01682108
  • Cas Number : 1847-24-1
  • Molecular Weight : 475.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1847-24-1.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/1847-24-1.html
pictograms GHS07
hstatements H315-H317-H335
Availability details for
Quantity
  • Availability
  • Add to Cart