AMBH324A4E3A Ambeed, Inc.

Phenethyl 3-(3,4-dihydroxyphenyl)acrylate

  • MDL number MFCD00866470
  • Cas Number : 104594-70-9
  • Molecular Weight : 284.3100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/104594-70-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/104594-70-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart