AMBH324A4E40 Ambeed, Inc.

Cytidine 5'-(trihydrogen diphosphate),disodium salt

  • MDL number MFCD00084682
  • Cas Number : 54394-90-0
  • Molecular Weight : 447.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/54394-90-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/54394-90-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart