AMBH324A4E46 Ambeed, Inc.

2,3',4',5'-Tetrafluoro-4-((1s,4r)-4-propylcyclohexyl)-1,1'-biphenyl

  • MDL number MFCD22380673
  • Cas Number : 173837-35-9
  • Molecular Weight : 350.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/173837-35-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/173837-35-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart