AMBH324A4E50 Ambeed, Inc.

(3aR,4S,7R,7aS)-Hexahydro-4,7-methanoisobenzofuran-1,3-dione

  • MDL number MFCD11046554
  • Cas Number : 14166-28-0
  • Molecular Weight : 166.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/14166-28-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14166-28-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart