AMBH324A4E53 Ambeed, Inc.

Boc-Arg HCl H2O

  • MDL number MFCD00065554
  • Cas Number : 204070-00-8
  • Molecular Weight : 328.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/204070-00-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/204070-00-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart