AMBH324A4E5A Ambeed, Inc.

(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

  • MDL number MFCD00005754
  • Cas Number : 958-09-8
  • Molecular Weight : 251.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/958-09-8.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/958-09-8.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart