AMBH324A4E5C Ambeed, Inc.

(trans,trans)-3,4-Difluorophenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate

  • MDL number MFCD30187304
  • Cas Number : 94840-77-4
  • Molecular Weight : 364.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/94840-77-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/94840-77-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart