AMBH324A4E60 Ambeed, Inc.

Miglitol

  • MDL number MFCD28142869
  • Cas Number : 72432-03-2
  • Molecular Weight : 207.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/72432-03-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/72432-03-2.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart