AMBH324A4E63 Ambeed, Inc.

Z-D-Asp(OtBu)-OH

  • MDL number MFCD00237335
  • Cas Number : 71449-08-6
  • Molecular Weight : 323.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/71449-08-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/71449-08-6.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart