AMBH324A4E64 Ambeed, Inc.

3'-O-tert-Butyldimethylsilyl-5'-O-DMT-N2-isobutyrylguanosine 2'-CE phosphoramidite

  • MDL number MFCD30475539
  • Cas Number : 1445905-51-0
  • Molecular Weight : 970.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1445905-51-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1445905-51-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart