AMBH324A4E73 Ambeed, Inc.

(S)-4-Amino-2-(((benzyloxy)carbonyl)amino)butanoic acid

  • MDL number MFCD00077904
  • Cas Number : 62234-40-6
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/62234-40-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/62234-40-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart