AMBH324A4E7C Ambeed, Inc.

Sodium deoxycholate

  • MDL number MFCD00064139
  • Cas Number : 302-95-4
  • Molecular Weight : 414.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/302-95-4.html

Safety Information

signalword Warning
pcodes P261
MSDS https://www.ambeed.com/products/302-95-4.html
pictograms GHS07
hstatements H302-H335
Availability details for
Quantity
  • Availability
  • Add to Cart