AMBH324A4E8E Ambeed, Inc.

Huperzine A

  • MDL number MFCD01714949
  • Cas Number : 102518-79-6
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/102518-79-6.html

Safety Information

signalword Danger
pcodes P260-P264-P280-P284-P301+P310-P302+P350
MSDS https://www.ambeed.com/products/102518-79-6.html
pictograms GHS06
hstatements H300-H310-H315-H319-H330-H335
Availability details for
Quantity
  • Availability
  • Add to Cart