AMBH324A4E96 Ambeed, Inc.

Halofuginone

  • MDL number MFCD09834143
  • Cas Number : 55837-20-2
  • Molecular Weight : 414.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/55837-20-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/55837-20-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart