AMBH324A4E9E Ambeed, Inc.

4-(trans-4-Propylcyclohexyl)cyclohexanone

  • MDL number MFCD04038797
  • Cas Number : 82832-73-3
  • Molecular Weight : 222.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/82832-73-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/82832-73-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart