AMBH324A4EA8 Ambeed, Inc.

Piceatannol

  • MDL number MFCD00221715
  • Cas Number : 10083-24-6
  • Molecular Weight : 244.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/10083-24-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10083-24-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart