AMBH324A4EB2 Ambeed, Inc.

(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate(relative)

  • MDL number MFCD00081070
  • Cas Number : 7534-94-3
  • Molecular Weight : 222.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7534-94-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/7534-94-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart