AMBH324A4EBE Ambeed, Inc.

(1R)-(-)-(10-Camphorsulfonyl)oxaziridine

  • MDL number MFCD00075428
  • Cas Number : 104372-31-8
  • Molecular Weight : 229.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/104372-31-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/104372-31-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart