AMBH324A4EC3 Ambeed, Inc.

Cinacalcet hydrochloride

  • MDL number MFCD08067750
  • Cas Number : 364782-34-3
  • Molecular Weight : 393.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/364782-34-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/364782-34-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart