AMBH324A4EC6 Ambeed, Inc.

Cephalosporin C zinc salt

  • MDL number MFCD00153937
  • Cas Number : 59143-60-1
  • Molecular Weight : 478.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/59143-60-1.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/59143-60-1.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart