AMBH324A4ECC Ambeed, Inc.

(E)-3-(p-Tolyl)acrylaldehyde

  • MDL number MFCD08460295
  • Cas Number : 56578-35-9
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/56578-35-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/56578-35-9.html
pictograms GHS07
hstatements H302-H315-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart