AMBH324A4ECF Ambeed, Inc.

H-Lys-OEt.2HCl

  • MDL number MFCD00039068
  • Cas Number : 3844-53-9
  • Molecular Weight : 210.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3844-53-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3844-53-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart