AMBH324A4ED4 Ambeed, Inc.

(R)-Tetrahydrofuran-3-amine 4-methylbenzenesulfonate

  • MDL number MFCD00040622
  • Cas Number : 111769-27-8
  • Molecular Weight : 259.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/111769-27-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/111769-27-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart