AMBH324A4EDD Ambeed, Inc.

(S)-tert-Butyl 1-methyl-2-oxoimidazolidine-4-carboxylate

  • MDL number MFCD09952527
  • Cas Number : 83056-79-5
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/83056-79-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/83056-79-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart