AMBH324A4EDF Ambeed, Inc.

(1S,2S)-2-Aminocyclohexanol hydrochloride

  • MDL number MFCD09259963
  • Cas Number : 13374-30-6
  • Molecular Weight : 151.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13374-30-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13374-30-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart