AMBH324A4EE2 Ambeed, Inc.

(2S,3S)-4-((4-Chlorophenyl)amino)-2,3-dihydroxy-4-oxobutanoic acid

  • MDL number MFCD00070634
  • Cas Number : 46834-56-4
  • Molecular Weight : 259.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/46834-56-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/46834-56-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart