AMBH324A4EE6 Ambeed, Inc.

trans-Methyl 4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate

  • MDL number MFCD17015041
  • Cas Number : 146307-51-9
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/146307-51-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/146307-51-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart