AMBH324A4EEC Ambeed, Inc.

(S)-3-(tert-Butyl(2-(2,4-difluorophenyl)-2-hydroxyethyl)amino)propanenitrile

  • MDL number MFCD18632987
  • Cas Number : 782482-51-3
  • Molecular Weight : 282.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/782482-51-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/782482-51-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart