AMBH324A4EF7 Ambeed, Inc.

(S)-1-(2-Chloroacetyl)pyrrolidine-2-carbonitrile

  • MDL number MFCD08689902
  • Cas Number : 207557-35-5
  • Molecular Weight : 172.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/207557-35-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/207557-35-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart