AMBH324A4EF8 Ambeed, Inc.

4-Propylphenyl 4-(trans-4-propylcyclohexyl)benzoate

  • MDL number MFCD01941025
  • Cas Number : 72928-02-0
  • Molecular Weight : 364.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/72928-02-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/72928-02-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart