AMBH324A4EFE Ambeed, Inc.

H-Hyp(tBu)-OH

  • MDL number MFCD00153445
  • Cas Number : 79775-07-8
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79775-07-8.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/79775-07-8.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart