AMBH324A4F03 Ambeed, Inc.

(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane)

  • MDL number MFCD22421614
  • Cas Number : 95756-62-0
  • Molecular Weight : 252.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/95756-62-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95756-62-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart