AMBH324A4F08 Ambeed, Inc.

tert-Butyl ((3R,4R)-4-aminotetrahydro-2H-pyran-3-yl)carbamate

  • MDL number MFCD19105154
  • Cas Number : 1240390-36-6
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1240390-36-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1240390-36-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart