AMBH324A4F0D Ambeed, Inc.

Tirofiban

  • MDL number MFCD05237246
  • Cas Number : 144494-65-5
  • Molecular Weight : 440.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/144494-65-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/144494-65-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart